Assay Details
Gene Symbol: ELAVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645363.1 | XP_023501131.1 | 721 | 70 | ||
XM_023645364.1 | XP_023501132.1 | 719 | 70 | |||
XM_023645365.1 | XP_023501133.1 | 718 | 70 | |||
XM_023645366.1 | XP_023501134.1 | 721 | 70 | |||
XM_023645367.1 | XP_023501135.1 | 718 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
