Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645374.1 | XP_023501142.1 | 603 | 96 | ||
XM_023645376.1 | XP_023501144.1 | 528 | 96 | |||
XM_023645377.1 | XP_023501145.1 | 603 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
