Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645374.1 | XP_023501142.1 | 755 | 92 | ||
XM_023645376.1 | XP_023501144.1 | 680 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
