Assay Details
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489017.6 | XP_001489067.1 | 979 | 63 | ||
XM_023645591.1 | XP_023501359.1 | 840 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
