Assay Details
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639026.1 | XP_023494794.1 | 929 | 73 | ||
XM_023639027.1 | XP_023494795.1 | 929 | 73 | |||
XM_023639028.1 | XP_023494796.1 | 906 | 73 | |||
XM_023639029.1 | XP_023494797.1 | 751 | 73 | |||
XM_023639030.1 | XP_023494798.1 | 929 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
