Assay Details
Gene Symbol: NACAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639024.1 | XP_023494792.1 | 7597 | 89 | ||
XM_023639025.1 | XP_023494793.1 | 7448 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
