Assay Details
Gene Symbol: RAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496097.5 | XP_001496147.4 | 483 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
