Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639101.1 | XP_023494869.1 | 983 | 70 | ||
XM_023639102.1 | XP_023494870.1 | 984 | 70 | |||
XM_023639103.1 | XP_023494871.1 | 636 | 70 | |||
XM_023639104.1 | XP_023494872.1 | 1846 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
