Assay Details
Gene Symbol: TMBIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609215.3 | XP_005609272.1 | 634 | 57 | ||
XM_023639186.1 | XP_023494954.1 | 593 | 57 | |||
XM_023639187.1 | XP_023494955.1 | 500 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
