Assay Details
Gene Symbol: HEPACAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609218.3 | XP_005609275.2 | 521 | 102 | ||
XM_014739195.2 | XP_014594681.1 | 520 | 102 | |||
XM_023639199.1 | XP_023494967.1 | 518 | 102 | |||
XM_023639200.1 | XP_023494968.1 | 518 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
