Assay Details
Gene Symbol: UMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739041.2 | XP_014594527.1 | 381 | 94 | ||
XM_014739042.2 | XP_014594528.1 | 349 | 94 | |||
XM_023639264.1 | XP_023495032.1 | 1678 | 94 | |||
XM_023639265.1 | XP_023495033.1 | 2336 | 94 | |||
XM_023639266.1 | XP_023495034.1 | 1710 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
