Assay Details
Gene Symbol: AGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739191.2 | XP_014594677.1 | 318 | 107 | ||
XM_014739192.2 | XP_014594678.1 | 330 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
