Assay Details
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609250.3 | XP_005609307.2 | 2500 | 75 | ||
XM_014739175.2 | XP_014594661.1 | 2388 | 75 | |||
XM_023639342.1 | XP_023495110.1 | 2575 | 75 | |||
XM_023639343.1 | XP_023495111.1 | 2507 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
