Assay Details
Gene Symbol: UBE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501795.6 | XP_001501845.2 | 503 | 63 | ||
XM_005609363.3 | XP_005609420.1 | 503 | 63 | |||
XM_014739049.2 | XP_014594535.1 | 504 | 63 | |||
XM_014739050.2 | XP_014594536.1 | 508 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
