Assay Details
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023639830.1 | XP_023495598.1 | 2296 | 63 | ||
XM_023639832.1 | XP_023495600.1 | 2200 | 63 | |||
XM_023639833.1 | XP_023495601.1 | 2296 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
