Assay Details
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364867.4 | XP_003364915.1 | 351 | 79 | ||
XM_005609455.3 | XP_005609512.1 | 351 | 79 | |||
XM_014739332.2 | XP_014594818.1 | 352 | 79 | |||
XM_014739334.2 | XP_014594820.1 | 351 | 79 | |||
XM_023639848.1 | XP_023495616.1 | 352 | 79 | |||
XM_023639849.1 | XP_023495617.1 | 360 | 79 | |||
XM_023639850.1 | XP_023495618.1 | 350 | 79 | |||
XM_023639853.1 | XP_023495621.1 | 351 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
