Assay Details
Gene Symbol: TCAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609470.3 | XP_005609527.2 | 1025 | 94 | ||
XM_023639882.1 | XP_023495650.1 | 1023 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
