Assay Details
Gene Symbol: TCAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609471.3 | XP_005609528.1 | 1779 | 66 | ||
XM_005609472.3 | XP_005609529.1 | 1785 | 66 | |||
XM_005609473.3 | XP_005609530.1 | 1924 | 66 | |||
XM_014739308.2 | XP_014594794.2 | 2055 | 66 | |||
XM_023639883.1 | XP_023495651.1 | 2052 | 66 | |||
XM_023639885.1 | XP_023495653.1 | 1848 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
