Assay Details
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646079.1 | XP_023501847.1 | 468 | 71 | ||
XM_023646080.1 | XP_023501848.1 | 226 | 71 | |||
GenBank mRNA | JL616027.1 | - | 2 - 3 | 126 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
