Assay Details
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504972.4 | XP_001505022.1 | 1817 | 76 | ||
XM_014741712.2 | XP_014597198.1 | 1879 | 76 | |||
XM_023646166.1 | XP_023501934.1 | 1655 | 76 | |||
GenBank mRNA | JL621385.1 | - | 3 - 4 | 651 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
