Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504989.6 | XP_001505039.1 | 1838 | 89 | ||
XM_023646177.1 | XP_023501945.1 | 904 | 89 | |||
XM_023646178.1 | XP_023501946.1 | 765 | 89 | |||
XM_023646179.1 | XP_023501947.1 | 1971 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
