Assay Details
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005612221.3 | XP_005612278.1 | 364 | 119 | ||
XM_023646180.1 | XP_023501948.1 | 221 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
