Assay Details
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646564.1 | XP_023502332.1 | 338 | 78 | ||
XM_023646565.1 | XP_023502333.1 | 329 | 78 | |||
XM_023646566.1 | XP_023502334.1 | 319 | 78 | |||
XM_023646567.1 | XP_023502335.1 | 313 | 78 | |||
XM_023646568.1 | XP_023502336.1 | 329 | 78 | |||
XM_023646569.1 | XP_023502337.1 | 327 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
