Assay Details
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646210.1 | XP_023501978.1 | 743 | 67 | ||
XM_023646211.1 | XP_023501979.1 | 585 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
