Assay Details
Gene Symbol: PRRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646218.1 | XP_023501986.1 | 2158 | 128 | ||
XM_023646219.1 | XP_023501987.1 | 2186 | 128 | |||
XM_023646220.1 | XP_023501988.1 | 528 | 128 | |||
XM_023646221.1 | XP_023501989.1 | 2153 | 128 | |||
XM_023646222.1 | XP_023501990.1 | 2182 | 128 | |||
XM_023646223.1 | XP_023501991.1 | 523 | 128 | |||
XM_023646224.1 | XP_023501992.1 | 2140 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
