Assay Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014742033.2 | XP_014597519.1 | 324 | 60 | ||
XM_023646696.1 | XP_023502464.1 | 2086 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
