Assay Details
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646726.1 | XP_023502494.1 | 1093 | 71 | ||
XM_023646727.1 | XP_023502495.1 | 1093 | 71 | |||
XM_023646728.1 | XP_023502496.1 | 1151 | 71 | |||
XM_023646729.1 | XP_023502497.1 | 1412 | 71 | |||
XM_023646730.1 | XP_023502498.1 | 2518 | 71 | |||
XM_023646731.1 | XP_023502499.1 | 1093 | 71 | |||
XM_023646732.1 | XP_023502500.1 | 814 | 71 | |||
XM_023646733.1 | XP_023502501.1 | 1151 | 71 | |||
XM_023646734.1 | XP_023502502.1 | 1408 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
