Assay Details
Gene Symbol: PPM1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646804.1 | XP_023502572.1 | 455 | 57 | ||
XM_023646805.1 | XP_023502573.1 | 408 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
