Assay Details
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496455.4 | XP_001496505.1 | 6152 | 71 | ||
XM_005612410.3 | XP_005612467.1 | 5764 | 71 | |||
XM_005612411.3 | XP_005612468.1 | 5625 | 71 | |||
XM_014742027.2 | XP_014597513.1 | 5540 | 71 | |||
XM_023646828.1 | XP_023502596.1 | 6358 | 71 | |||
XM_023646829.1 | XP_023502597.1 | 6087 | 71 | |||
XM_023646830.1 | XP_023502598.1 | 9073 | 71 | |||
XM_023646831.1 | XP_023502599.1 | 5519 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
