Assay Details
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499569.5 | XP_001499619.1 | 237 | 85 | ||
XM_014742119.2 | XP_014597605.1 | 302 | 85 | |||
XM_014742120.2 | XP_014597606.1 | 295 | 85 | |||
XM_014742121.2 | XP_014597607.1 | 293 | 85 | |||
XM_014742122.2 | XP_014597608.1 | 307 | 85 | |||
XM_023648132.1 | XP_023503900.1 | 292 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
