Assay Details
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500874.4 | XP_001500924.1 | 776 | 74 | ||
XM_005612494.3 | XP_005612551.1 | 811 | 74 | |||
XM_005612495.3 | XP_005612552.1 | 775 | 74 | |||
XM_014742069.2 | XP_014597555.1 | 1126 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
