Assay Details
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005612512.3 | XP_005612569.2 | 302 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005612512.3 transcripts.
Set Membership: |
![]() ![]() |
