Assay Details
Gene Symbol: C8H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488389.5 | XP_001488439.3 | 1006 | 114 | ||
XM_005612552.3 | XP_005612609.2 | 1000 | 114 |

More Information
Set Membership: |
![]() ![]() |
