Assay Details
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488554.5 | XP_001488604.1 | 1535 | 63 | ||
XM_023647023.1 | XP_023502791.1 | 1065 | 63 | |||
XM_023647024.1 | XP_023502792.1 | 885 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
