Assay Details
Gene Symbol: VSIG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489301.4 | XP_001489351.1 | 969 | 61 | ||
XM_005612570.3 | XP_005612627.1 | 688 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
