Assay Details
Gene Symbol: SDSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490133.5 | XP_001490183.1 | 639 | 59 | ||
XM_014727575.2 | XP_014583061.1 | 1280 | 59 | |||
XM_023647109.1 | XP_023502877.1 | 671 | 59 | |||
XM_023647110.1 | XP_023502878.1 | 1071 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
