Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496711.4 | XP_001496761.2 | 622 | 75 | ||
XM_023647219.1 | XP_023502987.1 | 491 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
