Assay Details
Gene Symbol: CCDC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005612621.3 | XP_005612678.1 | 1136 | 76 | ||
XM_014727553.2 | XP_014583039.1 | 1033 | 76 | |||
XM_023647246.1 | XP_023503014.1 | 1139 | 76 | |||
XM_023647247.1 | XP_023503015.1 | 1136 | 76 | |||
XM_023647248.1 | XP_023503016.1 | 1133 | 76 | |||
XM_023647249.1 | XP_023503017.1 | 1037 | 76 | |||
XM_023647250.1 | XP_023503018.1 | 1034 | 76 | |||
XM_023647251.1 | XP_023503019.1 | 1031 | 76 | |||
XM_023647252.1 | XP_023503020.1 | 1140 | 76 | |||
XM_023647253.1 | XP_023503021.1 | 1062 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
