Assay Details
Gene Symbol: CDK2AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647284.1 | XP_023503052.1 | 514 | 100 | ||
GenBank mRNA | JL628865.1 | - | 1 - 2 | 600 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
