Assay Details
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647282.1 | XP_023503050.1 | 768 | 74 | ||
XM_023647283.1 | XP_023503051.1 | 972 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
