Assay Details
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647354.1 | XP_023503122.1 | 744 | 53 | ||
XM_023647355.1 | XP_023503123.1 | 498 | 53 | |||
XM_023647356.1 | XP_023503124.1 | 557 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
