Assay Details
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647384.1 | XP_023503152.1 | 748 | 57 | ||
XM_023647385.1 | XP_023503153.1 | 745 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
