Assay Details
Gene Symbol: TWSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023647455.1 | XP_023503223.1 | 516 | 71 | ||
XM_023647456.1 | XP_023503224.1 | 521 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
