Assay Details
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648245.1 | XP_023504013.1 | 903 | 63 | ||
XM_023648247.1 | XP_023504015.1 | 948 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
