Assay Details
Gene Symbol: PSMA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495329.4 | XP_001495379.1 | 256 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
