Assay Details
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496712.6 | XP_001496762.4 | 231 | 59 | ||
XM_005612821.3 | XP_005612878.2 | 231 | 59 |

More Information
Set Membership: |
![]() ![]() |
