Assay Details
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496712.6 | XP_001496762.4 | 1842 | 76 | ||
XM_023647616.1 | XP_023503384.1 | 1852 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
