Assay Details
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498811.4 | XP_001498861.2 | 558 | 70 | ||
XM_005612883.3 | XP_005612940.2 | 763 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
