Assay Details
Gene Symbol: NARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488219.6 | XP_001488269.1 | 157 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488219.6 transcripts.
Set Membership: |
![]() ![]() |
