Assay Details
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005613001.3 | XP_005613058.1 | 869 | 75 | ||
XM_014727865.2 | XP_014583351.1 | 1702 | 75 | |||
XM_023648044.1 | XP_023503812.1 | 254 | 75 |
